missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK1 Monoclonal Antibody (8C5A7F10), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515631
This item is not returnable.
View return policy
Description
MA5-15631 targets CDC2 in indirect ELISA, FACS, IF, and WB applications and shows reactivity with Human samples. The MA5-15631 immunogen is purified recombinant fragment of CDC2 expressed in E. Coli. MA5-15631 detects CDC2 which has a predicted molecular weight of approximately 34kDa.
CDK1 or Cell Division Control protein 1 is essential for the completion of START, the controlling event in the cell cycle that is required to initiate mitosis. CDK1 is a catalytic subunit of a protein kinase complex, called the M-Phase Promoting Factor that induces entry into mitosis and is universal among eukaryotes. Phosphorylation of Bcl-2 in G2/M phase-arrested cells following photodynamic therapy with hypericin involves a CDK1-mediated signal and delays the onset of apoptosis. Therapeutic potential of CDK inhibitor NU2058 in androgen-independent prostate cancer has also been demonstrated.Specifications
CDK1 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P06493 | |
CDK1 | |
Purified recombinant fragment of CDC2 expressed in E. Coli. | |
RUO | |
983 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry, Western Blot | |
8C5A7F10 | |
Unconjugated | |
CDK1 | |
5363; ABL1; cdc; CDC2; cdc-2; cdc2 kinase; CDC28A; Cdc2a; cdc2-a; cdc2Dm; cdc2x1.1; CDCDm; CDK1; Cdk-1; cdk1.S; CDK1/CDC2; CDK1-A; cdk1-b; Cdk1-PA; CDKN1; cell cycle controller CDC2; cell cycle p34 CDC2 kinase protein; cell division control; cell division control protein 2 homolog; Cell division control protein 2 homolog 1; cell division control protein 2-A; cell division cycle 2 homolog A; cell division cycle 2 like; cell division cycle 2, G1 to S and G2 to M; Cell division cycle control protein 2; cell division cycle control protein 2a; cell division protein kinase 1; Cell division protein kinase 1-A; CG5363; CG5363-PA; cyclin dependent kinase; cyclin dependent kinase 1; cyclin-dependent kinase; Cyclin-dependent kinase 1; cyclin-dependent kinase 1 S homeolog; cyclin-dependent kinase 1-A; D2366; Dcdc2; dCdk1; DKFZp686L20222; Dm cdc2; Dmcdc2; DmCdk1; Dmel\CG5363; Dmel_CG5363; DNA polymerase delta catalytic subunit; DNA polymerase III; DNA-directed DNA polymerase delta POL3; group 4; HP | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |