missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK5 Monoclonal Antibody (4E4), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA517053
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
MA5-17053 targets CDK5 in indirect ELISA, FACS, ICC, IHC, IF and WB applications and shows reactivity with Human, Non-human primate, and Rat samples. The MA5-17053 immunogen is purified recombinant fragment of human CDK5 expressed in E. Coli. MA5-17053 detects CDK5 which has a predicted molecular weight of approximately 35kDa.
CDK5 is a member of the Cyclin-Dependent Kinase family that is most abundant in the mammalian brain. Active form of CDK5, which has also been called neuronal cdc2-like kinase, is a heterodimer of CDK5 and a 25 kDa protein which is derived proteolytically from a 35 kDa brain and neuron-specific protein and is essential for the kinase activity of CDK5. CDK5 has emerged as a crucial regulator of neuronal migration in the developing central nervous system. CDK5 phosphorylates a diverse list of substrates, implicating it in the regulation of a range of cellular processes - from adhesion and motility, to synaptic plasticity and drug addiction.![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Especificaciones
CDK5 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P49615, Q00535, Q03114 | |
CDK5 | |
Purified recombinant fragment of human CDK5 expressed in E. Coli. | |
RUO | |
1020, 12568, 140908 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
4E4 | |
Unconjugated | |
CDK5 | |
AW048668; Cdk5; CDK5 activator 1; CDK5-SV; CDKN5; Cell division protein kinase 5; CR6 protein kinase; Crk6; cyclin dependent kinase 5; cyclin-dependent kinase 5; cyclin-dependent-like kinase 5; LIS7; protein kinase CDK5 splicing; PSSALRE; Serine/threonine-protein kinase PSSALRE; tau protein kinase II catalytic subunit; TPKII catalytic subunit | |
Mouse | |
100 μL | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG1 |