missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEACAM5/CEACAM6 Monoclonal Antibody (MUS), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA117762
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This antibody recognizes native human CEACAM5, 6 protein expressed in transfected mammalian cells.
This gene encodes the light subunit of the ferritin protein. Ferritin is the major intracellular iron storage protein in prokaryotes and eukaryotes. It is composed of 24 subunits of the heavy and light ferritin chains. Variation in ferritin subunit composition may affect the rates of iron uptake and release in different tissues. A major function of ferritin is the storage of iron in a soluble and nontoxic state. Defects in this light chain ferritin gene are associated with several neurodegenerative diseases and hyperferritinemia-cataract syndrome. This gene has multiple pseudogenes.Especificaciones
CEACAM5/CEACAM6 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.2 | |
P06731, P40199 | |
CEACAM5, CEACAM6 | |
CEA partially purified from a perchloric acid extract from liver metastases of colonic tumors. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot | |
MUS | |
Unconjugated | |
CEACAM6 | |
Carcinoembryonic antigen; carcinoembryonic antigen related cell adhesion molecule 5; carcinoembryonic antigen related cell adhesion molecule 6; carcinoembryonic antigen-related cell adhesion molecule 5; Carcinoembryonic antigen-related cell adhesion molecule 6; carcinoembryonic antigen-related cell adhesion molecule 6 (non-specific cross reacting antigen); CD66c; CD66e; CEA; CEACAM5; CEACAM6; CEAL; Cluster of Differentiation 66c; meconium antigen 100; NCA; Non-specific crossreacting antigen; Normal cross-reacting antigen | |
Mouse | |
Protein G | |
RUO | |
1048, 4680 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |