Learn More
LABPLAS Cellulose Sani-Sponge with Dey-Engley Neutralizing Broth
Cellulose sponge for sampling surfaces, with buffer to neutralize disinfectant.
Marca: LABPLAS KSS-67115-DE
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- Cellulose Sani-Sponge
- Saturated with Dey-Engley neutralizing broth
- Dimensions: 160 x 229 mm
- Holds: 600 mL
- Gloves included with kit
Especificaciones
Hygiene Sponge Sampling Kit | |
Neutralizers | |
Sterile | |
Gloves |