missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ChAT Monoclonal Antibody (CL3173), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA531383
Detalles adicionales : Peso : 0.01000kg
Descripción
Immunogen sequence: GLFSSYRLPG HTQDTLVAQN SSIMPEPEHV IVACCNQFFV LDVVINFRRL SEGDLFTQLR KIVKMASNED ERLPPIGLLT SDGRSEWAEA RTVLVKDSTN
ChAT catalyzes the reversible synthesis of acetylcholine (ACh) from acetyl CoA and choline at cholinergic synapses.Especificaciones
ChAT | |
Monoclonal | |
1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P28329, P32738, Q03059 | |
CHAT | |
Recombinant protein corresponding to Human CHAT | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2b |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
CL3173 | |
Unconjugated | |
CHAT | |
acetyl CoA; acetyl CoA:choline O-acetyltransferase; B230380D24Rik; CHAT; CHOACTASE; choline acetylase; choline acetyltransferase; choline acetyltransferase (EC 2.3.1.6); choline acetyltransferase long variant; choline acetyltransferase short variant; choline O-acetyltransferase; CMS1A; CMS1A2; CMS6; LOW QUALITY PROTEIN: choline O-acetyltransferase; N-type choline acetyltransferase | |
Mouse | |
Protein A | |
RUO | |
1103, 12647, 290567 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |