Learn More
Tocris Bioscience™ CIM 0216
Selective TRPM3 agonist
Marca: Tocris Bioscience™ 5521/10
Detalles adicionales : CAS : 1031496-06-6 Peso : 0.00970kg
Descripción
CIM 0216 is a selective TRPM3 agonist. Exhibits selectivity for TRPM3 over TRPM1, TRPM2 and TRPM4-8. Stimulates insulin release from pancreatic islet cells in vitro. Evokes nocifensive behavior in mice.Especificaciones
3,4-Dihydro-N-(5-methyl-3-isoxazolyl)-α-phenyl-1(2H)-quinolineacetamide | |
1031496-06-6 | |
10 mg |