Learn More
Julabo&trade F Series Refrigerated Heating Circulating Baths with PID Sensor
Marca:  Julabo&trade 9116612
Código de producto. 11940218
Certificados
Ofertas especiales
Ofertas especiales
Título del producto
Julabo&trade F Series Refrigerated Heating Circulating Baths with PID Sensor > Range: -20 to 100deg.C; Pump Flow Rate: 15L/min.; Capacity: 4.5L; Cooling Capacity: