missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin 13 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA124420
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
PA1-24420 detects Claudin 13 in mouse samples. PA1-24420 has been successfully used in Western Blot procedures. PA1-24420 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of mouse Cldn13. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
CLDN13 is a member of claudin family. It is a four-transmembrane protein with WWCC motif, defined by W-X(17-22)-W-X(2)-C-X(8-10)-C.Especificaciones
Claudin 13 | |
Polyclonal | |
Unconjugated | |
Q9Z0S4 | |
CLDN13 | |
Synthetic peptide region of human Cldn13. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Western Blot | |
1 mg/mL | |
Cldn13 | |
2500004B14Rik, MGC144291, MGC144292 | |
Rabbit | |
Protein A | |
RUO | |
57255 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |