Learn More
Alpha Packaging™ Bote de vidrio transparente
Use the Alpha Packaging™ Clear Laboratory Glassware Pot as a fundamental piece of laboratory glassware. The autoclavable clear glass is breakproof and chemically and thermally shockproof.
Marca: Alpha Packaging™ 231258
Detalles adicionales : Peso : 7.00000kg
Descripción
- Clear borosilicate glass
- Break-resistant
- Chemically and thermally shockproof;
- In compliance with EP, USP and JP guidelines
- Autoclavable
Especificaciones
Contenedor | |
Vidrio | |
DIN 32 |
Conjunto de 126 | |
30 ml |