Learn More
Witeg™ Color Comparing Tubes Acc. to Eggertz
For the colorimetric estimation of carbon and manganese in steel.
Marca: Witeg™ 2 149 050
Detalles adicionales : Peso : 0.06400kg
Descripción
- With ST 12/21 stopper
- Capacity of 50 mL
- Manufactured from borosilicate glass
Especificaciones
50 mL | |
Stopper | |
ST 12/21 |
Borosilicate Glass | |
Colorimetric estimation |