missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement C3a Antibody (K13/16-5.7), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Bio-Techne NBP1-05122G
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Complement C3a Monoclonal specifically detects Complement C3a in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Especificaciones
Complement C3a | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
C3 | |
Human C3a | |
0.1 mL | |
Immunology, Innate Immunity, Neuronal Cell Markers, Neuroscience | |
718 | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
K13/16-5.7 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
AHUS5, ARMD9, ASP, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, complement C3, complement component 3, complement component C3, CPAMD1acylation-stimulating protein cleavage product | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Specific for human C3, C3a and C3a (desArg) and does not cross-react with C4a or C5a. | |
Store at 4C in the dark. | |
IgG1 κ |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido