Learn More
Invitrogen™ Complement C3d Monoclonal Antibody (053A-1149.3.1.4 (1003))

Mouse Monoclonal Antibody
Marca: Invitrogen™ MA183965
Descripción
Mouse anti Human C3d antibody, clone 053A-1149.3.1.4 recognizes human complement component 3d (C3d), a approximately 33 kDa polypeptide fragment generated over the course of complement activation where complement 3 (C3) convertases cleave C3 to C3b, which is further degraded into iC3b and C3dg/C3d.
C3 is a key component of the complement system since classical and alternative activation pathways merge at the C3 activation step when C3 is split into C3a and C3b. The molecular mass of C3 is 185 kDa and it consists of two chains (110 kDa and 75 kDa) held together by disulfide bonds.
Especificaciones
| Complement C3d | |
| Monoclonal | |
| 1.13 mg/mL | |
| BBS with 0.1% sodium azide; pH 8.4 | |
| P01024 | |
| C3 | |
| Purified C3d from human plasma. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Western Blot | |
| 053A-1149.3.1.4 (1003) | |
| Unconjugated | |
| C3 | |
| Acylation stimulating protein; Acylation stimulating protein cleavage product; acylation-stimulating protein cleavage product; AHUS5; AI255234; ARMD9; ASP; C3; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1; C3a; C3a anaphylatoxin; C3adesArg; C3b; C3bc; C3-beta-c; Cd3; chain fragment 2; Complement C3; Complement C3 alpha chain; Complement C3 beta chain; Complement C3b alpha' chain; Complement C3c alpha' chain fragment 1; Complement C3c alpha' chain fragment 2; Complement C3c alpha' Complement C3d fragment; Complement C3dg fragment; Complement C3f fragment; Complement C3g fragment; complement component 3; complement component C3a; complement component C3b; complement component C3d; CPAMD1; epididymis secretory sperm binding protein Li 62p; HEL-S-62p; HSE-MSF; Plp; prepro-C3; Unknown (protein for MGC:134562) | |
| Mouse | |
| Protein A | |
| RUO | |
| 718 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.