missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Connexin 50 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA511644
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This antibody is predicted to react with chicken based on sequence homology.
This gene encodes a transmembrane connexin protein that is necessary for lens growth and maturation of lens fiber cells. The encoded protein is a component of gap junction channels and functions in a calcium and pH-dependent manner. Mutations in this gene have been associated with zonular pulverulent cataracts, nuclear progressive cataracts, and cataract-microcornea syndrome.Especificaciones
Connexin 50 | |
Polyclonal | |
Unconjugated | |
GJA8 | |
Aey5; alpha 8 connexin; CAE; CAE1; cell surface glycoprotein; Cnx50; connexin 50; connexin-49; Connexin-50; CTRCT1; cx49; CX50; CZP1; gap junction alpha 8; gap junction alpha-8 protein; gap junction membrane channel protein alpha 8; gap junction membrane channel protein alpha-8; gap junction protein alpha 8; gap junction protein alpha 8 50kDa; gap junction protein, alpha 8; gap junction protein, alpha 8, 50kDa; GJA8; Lens fiber protein MP70; lens intrinsic membrane protein MP70; lens opacity 10; Lop10; MP38; MP64; MP70 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
14616, 2703 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1.6 mg/mL | |
PBS with 0.09% sodium azide | |
P28236, P48165 | |
GJA8 | |
KLH conjugated synthetic peptide between 402-433 amino acids from the C-terminal region of human Connexin 50 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |