missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COR 170, Tocris Bioscience™
Click to view available options
Cantidad:
10 mg
50 mg
Descripción
Selective inverse agonist of CB2 receptors (Ki values are 3.8 and >10,000 nM for CB2 and CB1, respectively).
Especificaciones
Nombre del producto químico o material | COR 170 |
Cantidad | 50 mg |
Formula Weight (peso de la fórmula) | 468.63 |
Porcentaje de pureza | >99% |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'