Learn More
Corning™ 1536-Well Black Clear-Bottom Polystyrene Plates
Highest density microplates available for high throughput screening
Marca: Corning™ 3895BC
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Conform to standard microplate footprint and dimensions. 2μLwell microplate is a thinner plate with the length and width dimensions and microplate footprint meeting industry standards.
- For fluorescent and luminescent assays
- Total well volume of 10μL for round well plates and 12.8μL for flat bottom plates
- Recommended working volume of up to 8μL
- Round well bottoms for reduced air entrapment and improved CVs and Z factor
- Raised well bottoms for higher sensitivity
- Flood reservoir on four sides to reduce instrument contamination
- Flat well bottoms
Información de órdenes
Packaged in bags of 10
Especificaciones
Standard | |
Clear | |
1536 | |
Non-sterile | |
Flat Bottom (F-well) |
32 x 48 | |
Polystyrene | |
8 μL | |
Non-binding | |
For fluorescent homogeneous assay |