Learn More
Corning™ 1536-Well Black Clear-Bottom Polystyrene Plates
Highest density microplates available for high throughput screening
Marca:  Corning™ 3895BC
Código de producto. 11313735
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos »
Explore más ofertas especiales
Descripción
Descripción
Conform to standard microplate footprint and dimensions. 2μLwell microplate is a thinner plate with the length and width dimensions and microplate footprint meeting industry standards.
  • For fluorescent and luminescent assays
  • Total well volume of 10μL for round well plates and 12.8μL for flat bottom plates
  • Recommended working volume of up to 8μL
  • Round well bottoms for reduced air entrapment and improved CVs and Z factor
  • Raised well bottoms for higher sensitivity
  • Flood reservoir on four sides to reduce instrument contamination
  • Flat well bottoms
Información de órdenes
Packaged in bags of 10
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
Standard
Clear
1536
Non-sterile
Flat Bottom (F-well)
32 x 48
Polystyrene
8 μL
Non-binding
For fluorescent homogeneous assay