Learn More
Corning™ Gentest™ Pre-Coated PAMPA Plate System
GREENER_CHOICE
A 96-well permeable support system with a filter plate pre-coated with structured layers of phospholipids.
Marca:  Corning™ 353015
Código de producto. 15305517
  • 977.00€ / paquete de 5
Fecha estimada de envío: 24-01-2025
para ver el stock
Explore más ofertas especiales
Descripción
  • Stable, pre-coated PAMPA plate is highly reproducible
  • Improved assay predictability over traditional PAMPA
  • Reduced mass retention of compounds
  • Well-suited as a ranking tool for the assessment of compounds with passive transport mechanisms and for predicting in vivo drug permeability
  • Polystyrene construction