missing translation for 'onlineSavingsMsg'
Learn More

CRMP5 Monoclonal Antibody (CR-1), Invitrogen™

Rat Monoclonal Antibody

Marca:  Invitrogen MA3701

Código de producto. 11565782

  • 489.00€ / 100 microlitros

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Explore más ofertas especiales
Este artículo no se puede devolver. Vea la política de devoluciones

Descripción

Descripción

MA3-701 detects human, bovine, rat and C-terminal region of CRMP5. This antibody has not been shown to be reactive in mouse samples. MA3-701 has been successfully used in Western blot and immunohistochemical procedures. By Western blot, this antibody detects an ∽ 62 kDa protein corresponding to human CRMP5 from brain extract. The MA3-701 antigen is a purified protein from human brain. This antibody is specific for the C-terminal region, amino acids 369 - 564. Not reactive with other CRMP isoforms.

The collapsin response mediator protein (CRMP) family of proteins (also referred to as ULIP6) are a group of neurologic proteins that have been found to play key roles in growth cone guidance during neural development. CRMP5 has relatively low sequence homology with the other four members of the CRMP family. CRMP5 is expressed in the developing nervous system in an expression pattern that resembles CRMP2 and has a peak expression during the first postnatal week. Elevated levels of autoantibody to CRMP5 have been shown to be linked to paraneoplastic autoimmune optic neuritis and other paraneoplastic neoplasms. Auto-immunity against CRMP5 expression (predominantly the N-terminal epitopes) is also being investigated as an oncological marker in the respiratory system and in the thymus.

Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly

TRUSTED_SUSTAINABILITY
Especificaciones

Especificaciones

CRMP5
Monoclonal
Conc. Not Determined
ascites with 0.05% sodium azide
Q9BPU6, Q9JHU0
DPYSL5
CRMP5 protein purified from human brain.
RUO
100126171, 56896, 65208
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot
CR-1
Unconjugated
DPYSL5
collapsin response mediator protein 5; collapsin response mediator protein-5; CRAM; CRMP3-associated molecule; Crmp5; CRMP-5; dihydropyrimidinase like 5; dihydropyrimidinase-like 5; dihydropyrimidinase-related protein 5; Dpysl5; DRP-5; Ulip6; ULIP-6; ULIP6 protein; UNC33-like phosphoprotein 6
Rat
100 μL
Primary
Bovine, Human, Rat
Antibody
IgG1
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado