missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTRP9/C1qTNF9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-46834-25ul
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CTRP9/C1qTNF9 Polyclonal antibody specifically detects CTRP9/C1qTNF9 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Especificaciones
| CTRP9/C1qTNF9 | |
| Polyclonal | |
| Western Blot 0.04 - 0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| B2RNN3 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 338872 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| AQL1, C1q and tumor necrosis factor related protein 9, C1QTNF9AComplement C1q tumor necrosis factor-related protein 9, complement C1q tumor necrosis factor-related protein 9A, CTRP9, MGC48915 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: DEMWLQVTGGERFNGLFADEDDDTTF | |
| 25 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido