Learn More
ACROBiosystems Cynomolgus / Rhesus macaque B7-1 / CD80 Protein, Fc Tag
314.52€ - 2153.18€
Especificaciones
Peso molecular | 50.5 kDa |
---|---|
Fuente | Cynomolgus / Rhesus macaque B7-1, Fc Tag (CD0-C5252) is expressed from human 293 cells (HEK293). It contains AA Val 35 - Asn 242 (Accession # G7NXN7). In the region Val 35 - Asn 242, the AA sequence of Cynomolgus and Rhesus macaque B7-1 are homologus. |
Inmunógeno | Val 35 - Asn 242 |
Requisitos de almacenamiento | -20°C |
Alias de gen | CD80,B7,B7-1,B7.1,BB1,CD28LG,CD28LG1,LAB7 |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15876107
|
ACROBiosystems
CD0-C5252-100UG |
100 μg |
314.52€
100 microgramos |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
15886107
|
ACROBiosystems
CD0-C5252-1MG |
1 mg |
2153.18€
1 miligramo |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
B7-1 and B7-2, together with their receptors CD28 and CTLA-4, constitute one of the dominant co-stimulatory pathways that regulate T- and B-cell responses. Although both CTLA-4 and CD28 can bind to the same ligands, CTLA-4 binds to B7-1 and B7-2 with a 20 - 100 fold higher affinity than CD28 and is involved in the down regulation of the immune response.B-lymphocyte activation antigen B7-1 (referred to as B7) also known as cluster of Differentiation 80 (CD80), is a member of cell surface immunoglobulin superfamily and is expressed on activated B cells, activated T cells, macrophages and dendritic cells. It is the ligand for two different proteins on the T cell surface: CD28 (for autoregulation and intercellular association) and CTLA-4 (for attenuation of regulation and cellular disassociation). CD80 works in tandem with CD86 to prime T cells. CD80 plays a role in induction of innate immune responses by activating NF-κB-signaling pathway in macrophages. CD80 is thus regarded as promising therapeutic targets for autoimmune diseases and various carcinomas.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 50.5 kDa. The protein migrates as 60-90 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.Especificaciones
50.5 kDa | |
Val 35 - Asn 242 | |
CD80,B7,B7-1,B7.1,BB1,CD28LG,CD28LG1,LAB7 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by reduced SDS-PAGE |
Cynomolgus / Rhesus macaque B7-1, Fc Tag (CD0-C5252) is expressed from human 293 cells (HEK293). It contains AA Val 35 - Asn 242 (Accession # G7NXN7). In the region Val 35 - Asn 242, the AA sequence of Cynomolgus and Rhesus macaque B7-1 are homologus. | |
-20°C | |
B7-1 | |
Cynomolgus Monkey, Rhesus Macaque | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Immune Checkpoint Proteins |