Learn More
ACROBiosystems Cynomolgus / Rhesus macaque PD-L1 / B7-H1 Protein, Fc Tag
423.38€ - 2661.24€
Especificaciones
Número de acceso | XP_014973154.1 |
---|---|
Peso molecular | 51.8 kDa |
Fuente | Cynomolgus / Rhesus macaque PD-L1, Fc Tag (PD1-C5253) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Arg 238 (Accession # F6VEW6-1). In the region Phe 19 - Arg 238, the AA sequence of Cynomolgus and Rhesus macaque PD-L1 are homologus. |
Inmunógeno | Phe 19 - Arg 238 |
Requisitos de almacenamiento | -20°C |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15887447
|
ACROBiosystems
PD1-C5253-100UG |
100 μg |
423.38€
100 microgramos |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
15897447
|
ACROBiosystems
PD1-C5253-1MG |
1 mg |
2661.24€
1 miligramo |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
Programmed cell death 1 ligand 1 (PDL1) is also known as B7-H, B7H1, MGC142294, MGC142296, PD-L1, PDCD1L1 and PDCD1LG1,which is a member of the growing B7 family of immune molecules and is involved in the regulation of cellular and humoral immune responses.PDL1 is a cell surface immunoglobulin superfamily with two Ig-like domains within the extracellular region and a short cytoplasmic domain. This protein is broadly expressed in the majority of peripheral tissues as well as hematopoietic cells. Interaction between PDL1 and its receptors belonging to the CD28 family of molecules provide both stimulatory and inhibitory signals in regulating T cell activation and tolerance. PDL1 may inhibit ongoing T-cell responses by inducing apoptosis and arresting cell-cycle progression.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 51.8 kDa. The protein migrates as 60-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
XP_014973154.1 | |
Cynomolgus / Rhesus macaque PD-L1, Fc Tag (PD1-C5253) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Arg 238 (Accession # F6VEW6-1). In the region Phe 19 - Arg 238, the AA sequence of Cynomolgus and Rhesus macaque PD-L1 are homologus. | |
-20°C | |
PD-L1 | |
Cynomolgus Monkey, Rhesus Macaque | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |
51.8 kDa | |
Phe 19 - Arg 238 | |
PD-L1,CD274,B7-H1,PDCD1L1,PDCD1LG1 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |