missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cys-mcMMAD, MedChemExpress
Click to view available options
Cantidad:
1 mg
Tamaño de la unidad:
1 miligramo
Description
Cys-mcMMAD is a drug-linker conjugate for ADC. MMAD is a potent tubulin inhibitor.
Specifications
Specifications
| Nombre del producto químico o material | Cys-mcMMAD |
| Color | Blanco |
| Notas de grado de pureza | Research |
| Fórmula molecular | C54H84N8O11S2 |
| Cantidad | 1 mg |
| SMILES | CC[C@H](C)[C@@H]([C@@H](CC(N1[C@@H](CCC1)[C@@H]([C@@H](C)C(N[C@H](C2=NC=CS2)CC3=CC=CC=C3)=O)OC)=O)OC)N(C([C@H](C(C)C)NC([C@H](C(C)C)N(C(CCCCCN4C(CC(SC[C@@H](C(O)=O)N)C4=O)=O)=O)C)=O)=O)C |
| Peso molecular (g/mol) | 1085.42 |
| Formula Weight (peso de la fórmula) | 1085.42 |
| Grado | Research |
| Forma física | Solid |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction