Learn More
CYTB Antibody - Azide and BSA Free, Novus Biologicals™
Código de producto. 18674522 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.02 mL
0.1 mL
Tamaño de la unidad:
0.02 ml
0.10 ml
18674522 0.02 mL 0.02 ml
18654272 0.1 mL 0.10 ml
2 options
Código de producto. 18674522
Marca: Novus Biologicals NBP2926390.02ml
Rabbit Polyclonal Antibody
CYTB Polyclonal antibody specifically detects CYTB in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno CYTB
Aplicaciones Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:100, Immunohistochemistry-Paraffin
Formulación PBS with 50% glycerol, pH7.3.
Alias de gen cytochrome b, MTCYB, MT-CYB mitochondrially encoded cytochrome b
Especie del huésped Rabbit
Inmunógeno A synthetic peptide corresponding to a sequence within amino acids 100-200 of human CYTB (YP_003024038.1). RGLYYGSFLYSETWNIGIILLLATMATAFMGYVLPWGQMSFWGATVITNLLSAIPYIGTDLVQWIWGGYSVDSPTLTRFFTFHFILPFIIAALATLHLLFL
Método de purificación Affinity purified
Cantidad 0.02 mL
Estado normativo RUO
Disciplina de investigación Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 4519
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at -20°C. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG