missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 6B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Especificaciones
Antígeno | Cytokeratin 6B |
---|---|
Concentración | 0.21 mg/mL |
Aplicaciones | Immunoprecipitation, Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) |
Clasificación | Polyclonal |
Conjugado | Unconjugated |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
16827274
|
Proteintech
17391-1-AP-20UL |
20 μL |
140.00€
20 microlitros |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
16817274
|
Proteintech
17391-1-AP-150UL |
150 μL |
399.00€
150 microlitros |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Descripción
The protein encoded by this gene is a member of the keratin gene family. The type II cytokeratins consist of basic or neutral proteins which are arranged in pairs of heterotypic keratin chains coexpressed during differentiation of simple and stratified epithelial tissues. As many as six of this type II cytokeratin (KRT6) have been identified; the multiplicity of the genes is attributed to successive gene duplication events. The genes are expressed with family members KRT16 and/or KRT17 in the filiform papillae of the tongue, the stratified epithelial lining of oral mucosa and esophagus, the outer root sheath of hair follicles, and the glandular epithelia. Mutations in these genes have been associated with pachyonychia congenita. The type II cytokeratins are clustered in a region of chromosome 12q12-q13.Especificaciones
Cytokeratin 6B | |
Immunoprecipitation, Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P04259, Q9Z331 | |
16688, 3854 | |
Cytokeratin 6B Fusion Protein Ag11288 | |
Primary | |
-20°C | |
KRT6B |
0.21 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CK 6B, CK6B, Cytokeratin 6B, K6B, keratin 6B, KRT6B, KRTL1, PC2, Type II keratin Kb10 | |
Krt6b | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |