Learn More
Thermo Scientific™ D-Glucose + D-Fructose + Sucrose
Product Code. 17847826 Shop All Thermo Scientific Products
Click to view available options
Cantidad:
40 x 25 unidades
Unit Size:
cada uno
This item is not returnable. View return policy
Product Code. 17847826
Brand: Thermo Scientific™ 984301
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
View alternative products
This item is not returnable. View return policy
D-Glucose + D-Fructose + Sucrose R1-R4 1000 tests R1 3 x 45mL R2-R4 3 x 13mL
D-Glucose + D-Fructose + Sucrose R1-R4 1000 tests R1 3 x 45mL R2-R4 3 x 13mL Ship and Store 2-8degC Do Not Freeze
D-Glucose + D-Fructose + Sucrose
  • R1-R4 1000 tests
  • R1 3 x 45mL
  • R2-R4 3 x 13mL
  • 40x25 Pcs
TRUSTED_SUSTAINABILITY
Specifications
Tipo D to Glucose + D to Fructose + Sucrose System Reagent
Tipo de muestra Food and Beverage Sample
Descripción Do Not Freeze
Detecta D-Glucose + D-Fructose + Sucrose
Nº de pruebas 1000
Tamaño de muestra R1-R4 1000 tests; R1 3 x 45 mL; R2-R4 3 x 13 mL
Cantidad 40 x 25 unidades
Contenido y almacenamiento 2°C to 8°C
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.