missing translation for 'onlineSavingsMsg'
Learn More

Clorhidrato de éster metílico de D-fenilalanina, 99 %, Thermo Scientific Chemicals

Catalog Number 11435670
Click to view available options
Cantidad:
5 g
25 g
Este artículo no se puede devolver. Vea la política de devoluciones
13033-84-6
C10H14ClNO2
215.68
MFCD00066112
SWVMLNPDTIFDDY-SBSPUUFOSA-N
d-phenylalanine methyl ester hydrochloride, h-d-phe-ome.hcl, d-phenylalanine methyl ester hcl, h-d-phe-ome hcl, r-methyl 2-amino-3-phenylpropanoate hydrochloride, d-phenylalanine, methyl ester, hydrochloride, methyl d-phenylalaninate hydrochloride, r-phenylalanine methyl ester hydrochloride, methyl 2r-2-amino-3-phenylpropanoate hydrochloride
11481330
methyl (2R)-2-amino-3-phenylpropanoate hydrochloride
Cl.COC(=O)[C@H](N)CC1=CC=CC=C1
Este artículo no se puede devolver. Vea la política de devoluciones
13033-84-6
C10H14ClNO2
215.68
MFCD00066112
SWVMLNPDTIFDDY-SBSPUUFOSA-N
d-phenylalanine methyl ester hydrochloride, h-d-phe-ome.hcl, d-phenylalanine methyl ester hcl, h-d-phe-ome hcl, r-methyl 2-amino-3-phenylpropanoate hydrochloride, d-phenylalanine, methyl ester, hydrochloride, methyl d-phenylalaninate hydrochloride, r-phenylalanine methyl ester hydrochloride, methyl 2r-2-amino-3-phenylpropanoate hydrochloride
11481330
methyl (2R)-2-amino-3-phenylpropanoate hydrochloride
Cl.COC(=O)[C@H](N)CC1=CC=CC=C1

Benzhydrylidene Aza-Glycinyl-D-Phenylalanine Methyl Ester was synthesized from D-phenylalanine methyl ester hydrochloride. Used in the conversion of amino acids into n-menthyloxycarbonyl alkyl ester derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
El éster metílico benzidrilideno aza-glicinil-D-fenilalanina se ha sintetizado a partir del clorhidrato de éster metílico de D-fenilalanina. Se utiliza en la conversión de aminoácidos en derivados del n-mentiloxicarbonil alquil éster.

Solubilidad
Soluble en etanol y metanol.

Notas
Almacenar en un lugar fresco. Mantenga el recipiente bien cerrado en un lugar seco y bien ventilado. Almacenar alejado de agentes oxidantes fuertes. Almacenar a 4 °C.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material D-Phenylalanine methyl ester hydrochloride
Punto de fusión 158°C to 162°C
Información de solubilidad Soluble in Ethanol and Methanol.
Formula Weight (peso de la fórmula) 215.68
Porcentaje de pureza 99%
Cantidad 5 g

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'