missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DbH Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA5122040
Este artículo no se puede devolver.
Vea la política de devoluciones
Descrizione
DbH Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
The protein encoded by this gene is an oxidoreductase belonging to the copper type II, ascorbate-dependent monooxygenase family. It is present in the synaptic vesicles of postganglionic sympathetic neurons and converts dopamine to norepinephrine. It exists in both soluble and membrane-bound forms, depending on the absence or presence, respectively, of a signal peptide.
Specifica
| DbH | |
| Polyclonal | |
| Unconjugated | |
| DBH | |
| Dbh; DBM; dopamine beta hydroxylase; Dopamine beta hydroxylase (dopamine beta-monooxygenase); dopamine beta-hydroxylase; dopamine beta-hydroxylase (dopamine beta-monooxygenase); dopamine beta-hydroxylase precursor; dopamine beta-hydroxylase precursor (EC 1.14.17.1); dopamine beta-monooxygenase; dopamine beta-monooxygenase precursor (EC 1.14.17.1); Dopamine-Beta-hydroxylase; DOPBHY; mixed function oxidase; Soluble dopamine beta-hydroxylase | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 1621 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P09172 | |
| DBH | |
| Recombinant protein Dopamine Beta Hydroxylase. The antigen corresponds to amino acid range 335-571 of the target protein. | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto