missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dectin 1 (soluble) Monoclonal Antibody (2A11), FITC, Invitrogen™
Rat Monoclonal Antibody
Marca: Invitrogen MA516479
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Rat anti Mouse Dectin-1 antibody, clone 2A11 recognizes murine beta-glucan receptor, also known as Dectin-1. Rat anti Mouse Dectin-1 antibody, clone 2A11 recognizes murine beta-glucan receptor, also known as Dectin-1.
CLEC7A, also known as dectin-1, is a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily and is predominantly expressed on myeloid cells. It is a small glycoprotein type II membrane receptor with an extracellular C-type lectin-like domain fold and a cytoplasmic domain with an immunoreceptor tyrosine-based activation motif (ITAM). CLEC7A functions as a pattern-recognition receptor that recognizes a variety of beta-1,3-linked and beta-1,6-linked glucans from fungi and plants, and in this way plays a role in innate immune response. Upon fungal exposure, CLEC7A activates Syk tyrosine kinase, triggering a massive oxidative burst through the formation of reactive oxygen species.Especificaciones
Dectin 1 (soluble) | |
Monoclonal | |
0.1 mg/mL | |
PBS with 1% BSA and 0.09% sodium azide | |
Q6QLQ4 | |
Clec7a | |
Dectin-1 transfected NIH3T3 cells and recombinant soluble Dectin-1 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2b |
Flow Cytometry | |
2A11 | |
FITC | |
Clec7a | |
Beta-glucan receptor; beta-GR; BGR; CANDF4; CD369; CLEC7A; Clec7b; CLECSF12; C-type (calcium dependent, carbohydrate recognition domain) lectin, superfamily member 12; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 12; C-type lectin domain family 7 member A; C-type lectin domain family 7, member A; C-type lectin superfamily member 12; DC-associated C-type lectin 1; DECTIN1; dectin-1; Dendritic cell-associated C-type lectin 1; dendritic cell-associated C-type lectin-1; lectin-like receptor 1; RGD1565140; SCARE2; UNQ539/PRO1082 | |
Rat | |
Protein G | |
RUO | |
56644 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |