Learn More
Sal sódica de ácido desoxirribonucleico, test de salmón, Thermo Scientific Chemicals
Catalog Number p-7043146
Click to view available options
Cantidad:
1 g
5 g
68938-01-2
C17H15BrFNO3
50000-100000
MFCD00130922
GXZCYECDYOBPGH-UHFFFAOYNA-N
SS-DNA
73904662
Deoxyribonucleic acid sodium salt
*
68938-01-2
C17H15BrFNO3
50000-100000
MFCD00130922
GXZCYECDYOBPGH-UHFFFAOYNA-N
SS-DNA
73904662
Deoxyribonucleic acid sodium salt
*
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Deoxyribonucleic acid sodium salt
Nombre de nota salmon testes
Color Blanco
Cantidad 1 g
Índice Merck 14,2907
Forma física Sólido