Learn More
TFA Dostmann Digital Thermometer with XL Probe
The digital thermometer with extra-long probe is ideal for an uncomplicated temperature measurement. The thermometer is HACCP and EN 13485 compliant. It’s also jet-proof and can be cleaned under running water after use.
Marca:  TFA Dostmann 30.1058.02
Código de producto. 17931662
  • 26.50€ / cada uno
Fecha estimada de envío: 22-04-2025
para ver el stock
Explore más ofertas especiales
Añadir a la cesta
Descripción
Descripción
This practical penetration probe thermometer from TFA with its extra-long probe is ideal for the uncomplicated temperature measurement of food. Simply stick or immerse the probe into what needs to be measured and within a very short time the device delivers accurate measurement results. The monitoring of the core temperature during the barbecuing or roasting of meats, the safe temperature check of baby food, the preparation food in the bakery, patisserie or in the catering industry, the control of the refrigerated counter at the butcher's or in the supermarket, stock checking or the inspection of incoming goods in the canteen kitchen or elsewhere in the food industry, these are all possible uses of the penetration probe thermometer. The thermometer is HACCP and EN 13485 compliant and well suited for professional use in industry, trade, commerce and laboratory. It's also jet-proof and can be cleaned under running water after use.
  • Thermometer with extra-long probe (300 mm)
  • Ideal for the control of food temperature
  • T90 measuring time about 8 sec.
  • Hold-, max.-min.-function with dynamic update
  • Automatic switch-off with long term measurement function
  • Compliant with HACCP + EN 13485, also for professional use
  • Accuracy ±0.5°C/1°F (from -10 to +100°C/+14 to +212°F), ±2°C/4°F (from +200 to +250°C/+392 to +482°F), otherwise ±1°C/2°F
  • Jet water-proof IP67
  • Registered TFA design
  • Can be calibrated
Especificaciones
Especificaciones
Plastic
Sí
LCD
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Ofertas especiales
Ofertas especiales
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
TFA Dostmann Digital Thermometer with XL Probe > Pantalla: LCD