Learn More
Gilac™ Dough Container
Dough Container is ideal for dough, pastries, chocolate molds storage, with rounded corners and bottom for easy cleaning.
Marca: Gilac™ G179421
Detalles adicionales : Peso : 1.27000kg
Descripción
- 600 x 400 dough container, ideal for dough, pastries, chocolate molds... storage.
- Stacking with and without lid, maximum stacking height capacity.
- Innovative design for even greater convenience:
- Rounded corners and bottom for easy cleaning.
- Vertical sides without inner edge avoids marking the dough.
- The entire surface of the container is usable.
- Completely flat bottom means dough detaches easily, no loss, no food deposit left.
- Optimised side reinforcements allows you to customise containers with your logo.
- Rounded outside corners guarantee a damage-free working environment.
- Translucent container facilitates the control of the contents.
- Allows you to monitor dough rising without lifting the container.
Especificaciones
Dough Container | |
White | |
4 gal. |
Polyethylene | |
600 x 400 x 90 mm | |
15 L |