missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DROSHA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA5122026
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Members of the ribonuclease III superfamily of double-stranded (ds) RNA-specific endoribonucleases participate in diverse RNA maturation and decay pathways in eukaryotic and prokaryotic cells (Fortin et al., 2002). The RNase III Drosha is the core nuclease that executes the initiation step of microRNA (miRNA) processing in the nucleus (Lee et al., 2003).Especificaciones
DROSHA | |
Polyclonal | |
Unconjugated | |
DROSHA | |
1110013A17Rik; AI874853; Drosha; drosha ribonuclease III; drosha, double-stranded RNA-specific endoribonuclease; drosha, ribonuclease type III; ethanol induced 2; ETOHI2; HSA242976; MGC115770; nuclear RNase III Drosha; OTTHUMP00000220910; OTTHUMP00000220911; OTTHUMP00000220912; OTTHUMP00000220913; p241; Protein Drosha; putative protein p241 which interacts with transcription factor Sp1; putative ribonuclease III; RANSE3L; RGD1307626; ribonuclease 3; Ribonuclease III; ribonuclease III, nuclear; ribonuclease type III, nuclear; RN3; RNase III; RNASE3L; RNASEN | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29102 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q9NRR4 | |
DROSHA | |
Ile1114-Glu1235 (Accession Q9NRR4), with N-terminal His Tag | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |