Learn More
Tocris Bioscience™ DSR 6434
Potent TLR7 agonist
Brand: Tocris Bioscience™ 4809/10
Additional Details : CAS : 1059070-10-8 Peso : 0.00970kg
Description
DSR 6434 is a potent TLR7 agonist (EC50 = 7.2 nM). Activates several immune effector cells in vitro. Suppresses metastasis in vivo and enhances efficacy of radiation therapy in a model of colorectal carcinoma.Specifications
DSR 6434 | |
6-Amino-2-(butylamino)-9-[[6-[2-(dimethylamino)ethoxy]-3-pyridinyl]methyl]-7,9-dihydro-8H-purin-8-one | |
Soluble to 50mM in 1eq. HCl and to 100mM in DMSO | |
Bioactive Small Molecule |
1059070-10-8 | |
10 mg | |
Solid |
Research Use Only. Not For Human or Veterinary Use