missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EIF2S1 Monoclonal Antibody (EIF2-alpha)
Mouse Monoclonal Antibody
Marca: Invitrogen™ AHO0802
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This antibody recognizes the alpha subunit of eukaryotic translation initiation factor 2 (EIF2-alpha). Recommended positive control are human Jurkat, CEM, and HeLa cells; mouse 3T3L1 cells; and rat PC-12 cells.
EIF2a (Eukaryotic translation initiation factor 2A) is a heterotrimer composed of three subunits (alpha, beta, and gamma). This translation initiation factor drives binding of initiator methionyl-tRNA to the 40S ribosome in an AUG-dependent manner to form the 43S pre-initiation complex. The polypeptide can be phosphorylated by related protein kinases activated in response to stress. Phosphorylated EIF2A inhibits EIF2B activity and prevents guanine nucleotide exchange.
Especificaciones
| EIF2S1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.4 | |
| P05198, P68101, Q6ZWX6 | |
| EIF2S1 | |
| Recombinant human eIF-2-alpha. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Monkey, Rat | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, RNA Immunoprecipitation, Immunocytochemistry | |
| EIF2-alpha | |
| Unconjugated | |
| EIF2S1 | |
| 0910001O23Rik; 2410026C18Rik; 35kDa; EIF2; EIF-2; eIF2 alpha; eIF2 alpha subunit; EIF2A; eIF-2A; eIF2alpha; EIF-2alpha; eIF-2-alpha; EIF2S1; eif2s1b; Eukaryotic translation initiation factor 2 subunit 1; eukaryotic translation initiation factor 2 subunit 1b; eukaryotic translation initiation factor 2 subunit alpha; eukaryotic translation initiation factor 2, subunit 1 alpha; eukaryotic translation initiation factor 2, subunit 1 alpha b; eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa; eukaryotic translation initiation factor 2A; fb19d01; translation initiation factor subunit; wu:fb19d01 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 13665, 1965, 54318 | |
| -20°C | |
| Liquid | |
| Responsibly packaged |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido