missing translation for 'onlineSavingsMsg'
Learn More

Erythropoietin alpha, human, Thermo Scientific Chemicals

Código de producto. 17750069
Click to view available options
Cantidad:
2 kU
Este artículo no se puede devolver. Vea la política de devoluciones
Este artículo no se puede devolver. Vea la política de devoluciones

Used in the study of red blood cell growth and differentiation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 11096-26-7
Fórmula molecular C22H22ClKN6O
Peso molecular (g/mol) 461.007
Clave InChI OXCMYAYHXIHQOA-UHFFFAOYSA-N
Sinónimo Recombinant Human Erythropoietin-^a,rHuEPO-^a
PubChem CID 11751549
Nombre IUPAC potassium;[2-butyl-5-chloro-3-[[4-[2-(1,2,3-triaza-4-azanidacyclopenta-2,5-dien-5-yl)phenyl]phenyl]methyl]imidazol-4-yl]methanol
SMILES CCCCC1=NC(=C(N1CC2=CC=C(C=C2)C3=CC=CC=C3C4=NN=N[N-]4)CO)Cl.[K+]

Especificaciones

Nombre del producto químico o material Erythropoietin alpha
Envase Frasco de plástico
Cantidad 2 kU
Formula Weight (peso de la fórmula) 21 kDa
Forma física Lyophilized Powder

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'