missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ESR1 Mouse anti-Human, Clone: OTI1B1, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Marca: Origene Technologies CF503759

Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Estrogen Receptors (ER) are members of the steroid/thyroid hormone receptor superfamily of nuclear receptors. The estrogen receptor is a ligand-activated transcription factor, that when bound to estrogen hormone, induces a conformational change that allows dimerization and binding to estrogen response elements (ERE) in DNA. When bound to EREs, ER can positively or negatively regulate gene transcription through the recruitment of coactivator or corepressor proteins. There are two different forms of the estrogen receptor, alpha and beta, encoded by separate genes (ESR1 and ESR2, respectively). Due to alternative RNA splicing, at least 4 estrogen receptor-alpha isoforms are known to exist (Isoform 1 (66 kDa), Isoform 2 (53 kDa), Isoform 3 (47 kDa), Isoform 4 (35 kDa)). Estrogen receptors are widely expressed in different tissue types and are essential for sexual development and reproductive function. They also play a role in other tissues such as bone. Estrogen receptors are involved in pathological processes including breast cancer, endometrial cancer, and osteoporosis.Especificaciones
ESR1 | |
Monoclonal | |
Unconjugated | |
ESR1 | |
ER, ESR, ESRA, ESTRR, Era, NR3A1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
2099 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1B1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P03372 | |
ESR1 | |
Full length human recombit protein of human ESR1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Corrección del contenido de un producto
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.
Título del producto
ESR1 Mouse anti-Human, Clone: OTI1B1, lyophilized, TrueMAB™
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido