missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ESR1(phospho S118) & ESR1 Protein Phosphorylation Antibody Pair, Abnova™
Antibody Pair
Brand: Abnova DP0157.1Set
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
This protein phosphorylation antibody pair set comes with two antibodies, one against the ESR1 protein, and the other against the specific S118 phosphorylated site of ESR1 for use in in situ Proximity Ligation Assay.Specifikationer
| ESR1 | |
| Unconjugated | |
| DKFZp686N23123/ER/ESR/ESRA/Era/NR3A1 | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Phospho-ESR1 S118 rabbit polyclonal antibody (20 ul) In PBS, 150 mM NaCl, pH 7.4 (0.02% sodium azide, 50% glycerol) 2. ESR1 mouse monoclonal antibody (40 ug) In 1x PBS, pH 7.2 *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| In situ PLA | |
| estrogen receptor 1 | |
| ESR1 | |
| RUO | |
| 2099 | |
| Human | |
| Ab Pair |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering