missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETS1 Monoclonal Antibody (1G11), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA512375
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
MA5-12375 targets Ets-1 in IHC (P) applications and shows reactivity with Human samples. The MA5-12375 immunogen is recombit oncoprotein corresponding to aa 226-454 of human Ets-1.
Ets-1 is a transcription factor involved in the regulation of genes required for extracellular matrix remodeling during metastasis of tumors. It is known to interact with the enhancer of urokinase like plasminogen activator, and promoters of stromelysin-1 and collagenase-1. It is absent from normal gastric mucosa but is expressed in 60% of gastric carcinoma and oral squamous cell carcinoma.Especificaciones
ETS1 | |
Monoclonal | |
Unconjugated | |
ETS1 | |
Avian erythroblastosis virus E26 (v-ets) oncogene homolog-1, ets protein, p54, protein C-ets-1, v-ets avian erythroblastosis virus E2 oncogene homolog 1, v-ets avian erythroblastosis virus E26 oncogene homolog 1 | |
Mouse | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Immunohistochemistry (Paraffin) | |
1G11 | |
tissue culture supernatant with 0.09% sodium azide | |
P14921 | |
ETS1 | |
Recombit oncoprotein corresponding to aa 226-454 of human Ets-1 | |
RUO | |
2113 | |
4° C | |
Liquid |