missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETV6 Monoclonal Antibody (BM3), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA191170
Detalles adicionales : Peso : 0.01000kg
Descripción
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes an ETS family transcription factor. The product of this gene contains two functional domains: a N-terminal pointed (PNT) domain that is involved in the protein-protein interactions with itself and other proteins, and a C-terminal DNA-binding domain. Gene knockout studies in mice suggest that it is required for hematopoiesis and Maintenance of the developing vascular network. This gene is known to be involved in a large number of chromosomal rearrangements associated with leukemia and congenital fibrosarcoma.Especificaciones
ETV6 | |
Monoclonal | |
1 mg/mL | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
P41212 | |
Etv6 | |
A BALB/c mouse was immunized with nuclei from Pokeweed mitogen stimulated human peripheral blood lymphocytes | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunoprecipitation, Western Blot | |
BM3 | |
Unconjugated | |
Etv6 | |
AW123102; AW557856; CGI-108; CSL4; Csl4p; ETS translocation variant 6; ETS variant 6; ets variant gene 6 (TEL oncogene); ETS-related protein Tel1; Etv6; hCsl4p; p13; RP11-452K12.9; SKI4; Ski4p; Tel; TEL/ABL; Tel1; TEL1 oncogene; THC5; Transcription factor ETV6 | |
Mouse | |
Protein A | |
RUO | |
2120 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |