Learn More
Univet™ Evolution Disposable Sights and Frames Package
The Evolution model is equipped with very light frames in different colors and with foldable temples to make it possible to put the product in the pocket after removing the disposable visor with anti-fog coating.
Marca: Univet™ MDU7003
Detalles adicionales : Peso : 0.08700kg
Descripción
- Contains 10 kits for a total of 50 sights and 10 frames
- Each kit consists of 5 interchangeable disposable sights and 1 colored frame
- Foldable
- Anti-fog and disposable lens
- Interchangeable visor
- Ultra-light weight
- 5 colorful frames
Especificaciones
Oval | |
None | |
10 Kits/Pk. | |
Reusable | |
Plastic | |
Not Autoclavable |
Scratch-resistant | |
Eyeshield | |
Polyester | |
Polyester | |
None |