Learn More
Thermo Scientific™ Espectrofotómetros Evolution™ One/One Plus UV-Vis
Los espectrofotómetros Thermo Scientific™ Evolution™ UV-Vis combinan un hardware fiable y versátil con el software Thermo Scientific™ Insight™ Pro fácil de usar y de aprender.Juntos, satisfarán las diversas necesidades que su organización requiere de una herramienta analítica.
Marca: Thermo Scientific™ 840-370100
Descripción
Para la investigación rutinaria y compleja y las aplicaciones avanzadas, incluida la conformidad actual, la serie de espectrofotómetros Thermo Scientific™ Evolution™ One/One Plus UV-Vis combina un hardware fiable y versátil y una amplia gama de accesorios con el software Thermo Scientific™ Insight™ Pro fácil de usar y de aprender.
Seminario web:Identificación de la agregación de proteínas mediante espectroscopía UV-Vis
For routine to research analysis, this instrument series offers real innovations that provide enhanced usability and performance - without added complexity. With straightforward software, cutting-edge instrumentation and a wide selection of accessories, the Evolution One and One Plus UV-Vis Spectrophometers work with you to perform experiments the way you want - for the results you need.
Optimized Engineering Delivers Ultimate Performance:- Double-beam geometry with fixed 1nm or variable spectral bandwidth options
- Application Focused Beam Geometry (AFBG) technology enables the best performance from microcells, solid samples, and fiber optic probes (Evolution One Plus Only)
- Xenon flash lamp requires no warm-up time allowing instant measurements
- Meet chemical, materials science, or biological sampling and measurement needs with a broad range of accessories
- Reduce training time: Optimized user interface with prompting software defines clear step-by-step workflow and is easy to teach and learn
- Prevent costly sampling and measurement errors: Create dedicated workflows for complex assay and transform multi-step workflows into one simplified, easy-to-follow method suitable for any level of user with Customized User Environment (CUE)
- Always current: Whether it is user inputs or application requirements, or new regulations, Insight Pro Software is continuously improving with quick upgrades to deliver greater capabilities and efficiencies for your lab’s needs
- Wide accessory selection with various temperature control options, sample changers, sippers and solid sampling devices complete your analysis
- Choice of Peltier Single Cell Holder or a Smart 8-Cell Peltier system for temperature control and sample monitoring from 0° to 100 °C
- Easy software integration gives you complete control of your measurements
Regulatory/Quality Control Environments:
To use the EvolutionOne/One Plus UV-Vis Spectrophotometers in a regulated or quality control environment, we offer a wide selection of compliance products and services to reduce implementation time and assure system performance.
- Evolution One or One Plus UV-Vis Spectrometer
- INSIGHT Pro with Security Software
- Validator for Insight Pro US Ltr or INTL A4 Paper Version
- Qualification Filter Kit for UV-Vis
- Evolution One or One Plus Spectrometer
- INSIGHT Pro with Security Software
- Validator for Insight Pro US Ltr or INTL A4 Paper Version
- USP and EP UV Standards Set
- USP and EP Visible Standards Set
Especificaciones
Evolution One Security and Validator Bundle includes: -Evolution One UV-Vis Spectrometer -INSIGHT Pro with Security Software -Validator for Insight Pro US Ltr Paper Version -Qualification Filter Kit for UV-Vis |
|
Evolution One | |
1 nm | |
Paquete de seguridad y validador: EE. UU |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.