Learn More
KARTELL Eye Wash Station
Click to view available options
:
Eye Wash Station
Descripción
- Emergency eye wash station moulded in strong high-impact PS.
- The directions are clearly printed on the support rack and the bottle can be filled with distilled water or other medicated liquid for ophthalmic use.
- Eye wash bottle art. 0038300, to be ordered separately, is made of PE provided with an eye cup with snap on cap, carefully designed to allow a quick and immediate flushing to remove foreign matters and prevent serious eye damage.
- The head has an air-vent valve to assure air refill of free space in the bottle thus preventing draw back of contaminated wash into the bottle.
- A perforated spray plate in the cup centre breaks up the stream to give the eye a gentle wash.
- A drain tube releases waste.
Especificaciones
Especificaciones
| Color | Blanco |
| Material | Polietileno |