missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F(ab')2-Donkey anti-Rat IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, FITC, Invitrogen™
F(ab')2-Donkey Polyclonal Secondary Antibody
Marca: Invitrogen A24544
Detalles adicionales : Peso : 0.01000kg
Descripción
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Especificaciones
Rat IgG (H+L) Highly Cross-Adsorbed | |
Polyclonal | |
FITC | |
Donkey | |
Affinity Chromatography | |
RUO | |
Rat | |
Liquid |
Immunocytochemistry | |
1.5 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.8 | |
Gamma Immunoglobins Heavy and Light chains | |
500 μg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
IgG |