missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ F(ab)-Goat anti-Rabbit IgG Fc Secondary Antibody, Alexa Fluor™ 647

Código de producto. 30019260
Change view
Click to view available options
Cantidad:
250 μg
Tamaño de la unidad:
250 microgramos
Código de producto. Cantidad unitSize
30019260 250 μg 250 microgramos
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 30019260

Marca: Invitrogen™ A66788

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

F(ab)-Goat Polyclonal Secondary Antibody

This F(ab)-Goat anti-Rabbit IgG Fc specific antibody product is a goat polyclonal antibody F(ab) fragment that is specific to the Fc region of rabbit IgG. The product has minimal to no cross-reactivity towards polyclonal mouse, rat, guinea pig, bovine, hamster, horse, sheep, human, monkey, donkey IgG, and chicken IgY as tested by direct ELISA. Cross-reactivity towards other immunoglobulins has not been tested. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. F(ab) antibody fragment offer several benefits over whole IgGs, due to their smaller size (~50 kD vs. ~150 kD) and the absence of the Fc region. The smaller size is expected to allow better penetration into tissue sections and improved staining in applications such as IHC. The lack of Fc region results in reduced nonspecific background staining when staining cells/ tissues with Fc receptors (such as immune cells) and other Fc interaction-related interference. Fc specific F(ab) fragments provide a range of multiplexing options beyond what is possible with whole IgGs for applications such as immunocytochemistry, immunohistochemistry, flow cytometry, and western blotting. Using conjugate solutions: Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment. This step will help eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining.

Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Rabbit IgG Fc
Aplicaciones Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Clasificación Polyclonal
Concentración 0.2 mg/mL
Conjugado Alexa Fluor 647
Formulación PBS with 5mM sodium azide; pH 7.5
Especie del huésped Goat
Inmunógeno Rabbit IgG Fc.
Método de purificación Size-exclusion chromatography
Cantidad 250 μg
Estado normativo RUO
Primario o secundario Secondary
Especies diana Rabbit
Contenido y almacenamiento 4°C, store in dark, DO NOT FREEZE!
Tipo de producto Secondary Antibody
Formulario Liquid
Isotype IgG F(ab)
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.