Learn More
R&D Systems™ Recombinant F. meningosepticum PNGase F Protein, CF
Effective removal of N-linked oligosaccharides from glycoproteins. Glycerol-free, suitable for downstream MS and HPLC. Get 50% off! Use Promo Code: PNGaseF50 Terms and Conditions
Marca:  R&D Systems™ 9109-GH-020
Encompass
Código de producto. 15740394
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos »
Descripción
Descripción
Measured by its ability to deglycosylate ribonuclease B under denatured conditions. >50% ribonuclease B (10 μg) is deglycosylated by 2.5 ng rFmPNGase F within 30 minutes, as measured under the described conditions.
Especificaciones
Especificaciones
P21163
M.W. (Observed): 34 kDa, reducing conditions; M.W. (theoretical): 36 kDa
E. coli-derived f. meningosepticum PNGase F protein Ala41-Asn354 with N-terminal Met and 6-His tag
<1.0 EU / 1 µg of the protein by the LAL method.
Measured by its ability to deglycosylate ribonuclease B under denatured conditions. >50% ribonuclease B (10μg) is deglycosylated by 2.5 ng rFmPNGase F within 30 minutes, as measured under the described conditions.
Yes
Supplied as a 0.2 µm filtered solution in Tris and NaCl.
20 µg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
PNGase F
Unconjugated
Certificados
Promociones
Promociones
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.