missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FANCD2 Monoclonal antibody specifically detects FANCD2 in Human samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Flow Cytometry, KnockDown
Specifications
Specifications
| Antígeno | FANCD2 |
| Aplicaciones | Western Blot, Immunohistochemistry, Flow Cytometry, KnockDown |
| Clasificación | Monoclonal |
| Clon | 1290D |
| Conjugado | PE |
| Dilución | Western Blot, Simple Western, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Knockout Validated |
| Formulación | PBS |
| Alias de gen | DKFZp762A223, FA4, FACD, FAD, FAD2, FA-D2, FADFAD2, FANCD, Fanconi anemia complementation group D2, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, FLJ23826, FPN1, HFE4, IREG1, Protein FACD2, SLC11A3 |
| Especie del huésped | Rabbit |
| Inmunógeno | E. coli-derived recombinant human FANCD2, Glu11-Glu230, Accession # Q9BXW9 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?