missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Feline IL-10 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marca: R&D Systems MAB736-SP
146.40 EUR hasta el 2025-03-28
Use el código promocional "25306" para aplicar la oferta.
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
IL-10 Monoclonal specifically detects IL-10 in Feline samples. It is validated for Western Blot, Neutralization.
Especificaciones
IL-10 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3586 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Western Blot, Neutralization | |
137706 | |
Western Blot 1 ug/mL, Neutralization 0.2-0.6 ug/mL | |
P55029 | |
IL10 | |
E. coli-derived recombinant feline IL-10 Ser19-Ile178 Accession # P55029 | |
25 μg | |
Primary | |
Detects feline IL-10 in direct ELISAs and Western blots. In direct ELISAs, 40% cross-reactivity with canine IL-10 is observed and no cross-reactivity with IL-10 from human, mouse, rat, Epstein-Barr virus, or human cytomegalovirus is observed. | |
Feline | |
Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Feline IL-10 Antibody, R&D Systems™ > 25μg; Unlabeled
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido