Descripción
Rotating locks minimize twisting and kinking of tubing during positioning. Tapered cone seals are reliable and uncomplicated. Facilitate single-use configurations that minimize downtime
Especificaciones
Especificaciones
| Certificaciones/conformidad | Non-DEHP;Non-Phthalate;RoHS |
| Diámetro interior (imperial) | 5/32 pulg |
| Material | Nailon |
| Cantidad de embalaje | 25/Pk. |
| Conexión | Female Luer x Hose Barb |
Always test the application to verify the performance of the interconnection.