Learn More
Ferritin Heavy Chain Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 2 publications
Marca: Novus Biologicals NBP1-06985
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Ferritin Heavy Chain Polyclonal specifically detects Ferritin Heavy Chain in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Peptide ELISA.For Research Use Only