Learn More
FGF-13 Antibody (298923), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Marca:  Novus Biologicals FAB1909D
Código de producto. 18588398
  • 601.00€ / 0.10 ml
Explore más ofertas especiales
Description
Description
FGF-13 Monoclonal antibody specifically detects FGF-13 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FGF-13
Monoclonal
DyLight 350
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Western Blot, Flow Cytometry, CyTOF
298923
Western Blot, Flow Cytometry, CyTOF-ready
FGF-13, FGF2Fibroblast growth factor homologous factor 2, FHF2FHF-2, fibroblast growth factor 13
E. coli-derived recombinant human FGF-13 isoform 1B, Ala2-Thr192, Accession # NP_378668
0.1 mL
Angiogenesis, Biologically Active Proteins, Neuroscience
2258
Store at 4°C in the dark.
IgG2a
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FGF-13 Antibody (298923), DyLight 350, Novus Biologicals™ > 0.1 mL; DyLight 350
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.