Learn More
Fisherbrand™ Baños de agua de control digital Isotemp™: modelo 202
Ideales para aplicaciones industriales, clínicas, farmacéuticas o biomédicas
Descripción
Ideal for industrial, clinical, pharmaceutical or biomedical applications
Microprocessor PID Control
- ±0.24°C uniformity, ±0.5°C accuracy and stability at 37°C
- Display shows °C or °F; easy to read from across the room
- Low profile and front-panel controls for easier access, safer operation
- Dual thermostats guard against thermal runaway
- Hinged acrylic or stainless-steel gable covers have fins that keep hands away from hot vapors
- Covers stay open at 90° or, with quick-release hinge, lift off completely for use with large glassware
- Heating elements will not burn out if bath accidentally runs dry
- Corrosion-resistant exterior with tough, easy-to-clean acrylic enamel coating
- Temperature-resistant plastic rim insulates, resists leaks, keeps its shape and won't dry out or crack like caulked seals
- Exterior stays cool even after hours of operation Deep-drawn, stainless-steel reservoirs and perforated sample trays prevent leaks, guard against rust and contamination
- Rounded reservoir corners and sample trays with finger holes allow for easy removal, make cleaning easier
- Choice of acrylic or stainless-steel covers
Información de órdenes
Additional voltages and models with analog controllers available; contact your Fisher Scientific Customer Service Representative for details.
Service Plan
Service agreement available.
Certifications
CSA approved.
Garantía
One year